Learn More
Medchemexpress LLC 4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one | 954373-94-5 | 95.9% | 335.98 | 500 MG
SDP
Catalog No. 5000400587
Encompass_Preferred
4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Drug intermediate for the synthesis of active compounds
  • For research use only
  • Not sold to patients
  • Solid, white to off-white appearance
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
Catalog No. 50-004-00587 Supplier Medchemexpress LLC Supplier No. HY22018500MG
4,5,6,7-Tetrafluoro-1-hydroxy-1,2-benziodoxole-3(1H)-one is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Drug intermediate for the synthesis of active compounds
  • For research use only
  • Not sold to patients
  • Solid, white to off-white appearance
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month