Learn More
Medchemexpress LLC 4,5,6,7-Tetrafluoro-L-tryptophan | 72120-72-0 | 99.99% | 276.19 | 100 MG
SDP
Catalog No. 5000386241
Encompass_Preferred
F4-Trp (4,5,6,7-Tetrafluoro-L-tryptophan) is a ligand that targets the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR). It shows promise for research in neurotransmission, addiction, and related conditions, including neurological diseases. It is relevant for research in neurological, eye or ear diseases, and membrane transporter/ion channel and neuronal signaling pathways.
  • Ligand targeting α3β4 nicotinic acetylcholine receptor
  • Useful for neurotransmission research
  • Applicable to addiction research
  • Potential for neurological disease studies
  • Inhibitor of nAChR
Catalog No. 50-003-86241 Supplier Medchemexpress LLC Supplier No. HY171746100MG
F4-Trp (4,5,6,7-Tetrafluoro-L-tryptophan) is a ligand that targets the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR). It shows promise for research in neurotransmission, addiction, and related conditions, including neurological diseases. It is relevant for research in neurological, eye or ear diseases, and membrane transporter/ion channel and neuronal signaling pathways.
  • Ligand targeting α3β4 nicotinic acetylcholine receptor
  • Useful for neurotransmission research
  • Applicable to addiction research
  • Potential for neurological disease studies
  • Inhibitor of nAChR