Learn More
Medchemexpress LLC 4,5,6,7-Tetrafluoro-L-tryptophan | 72120-72-0 | 99.99% | 276.19 | 25 MG
SDP
Catalog No. 5000383927
Encompass_Preferred
F4-Trp (4,5,6,7-Tetrafluoro-L-tryptophan) is a ligand that targets the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR). It is considered promising for research into neurotransmission, addiction, and related neurological diseases.
  • Ligand targeting the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR)
  • Promising for research of neurotransmission, addiction, and neurological diseases
  • High purity of 99.99%
Catalog No. 50-003-83927 Supplier Medchemexpress LLC Supplier No. 25MGHY171746
F4-Trp (4,5,6,7-Tetrafluoro-L-tryptophan) is a ligand that targets the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR). It is considered promising for research into neurotransmission, addiction, and related neurological diseases.
  • Ligand targeting the α3β4 nicotinic acetylcholine receptor (α3β4 nAChR)
  • Promising for research of neurotransmission, addiction, and neurological diseases
  • High purity of 99.99%