Learn More
Medchemexpress LLC 4,5,6,7-tetrahydro-N-[3-methoxy-4-(4-methyl-1H-imidazol-1-yl)phenyl]-4-phenyl-2-benzothiazolamine | 1172637-87-4 | 98.0% | 416.54 g/mol | C24H24N4OS | 5 MG
SDP
Catalog No. 5000214125
Encompass_Preferred
gamma-secretase modulator 1 is a small-molecule research reagent that modulates γ-secretase activity and is used to study amyloid-β production and Alzheimer's disease-related pathways. It is supplied as a white to off-white solid with reported purity of 98.0% and a molecular weight of 416.54 g/mol. For research use only; consult the product datasheet and SDS for storage and handling.
  • Modulates γ-secretase activity for amyloid-β research.
  • High purity (98.0%) suitable for analytical studies.
  • White to off-white solid form for ease of handling.
  • Limited solubility in DMSO (< 1 mg/mL).
  • Recommended storage: powder -20 °C (long term) or 4 °C (short term).
  • Product datasheet and SDS available for safety and handling.
Catalog No. 50-002-14125 Supplier Medchemexpress LLC Supplier No. HY100435MG
gamma-secretase modulator 1 is a small-molecule research reagent that modulates γ-secretase activity and is used to study amyloid-β production and Alzheimer's disease-related pathways. It is supplied as a white to off-white solid with reported purity of 98.0% and a molecular weight of 416.54 g/mol. For research use only; consult the product datasheet and SDS for storage and handling.
  • Modulates γ-secretase activity for amyloid-β research.
  • High purity (98.0%) suitable for analytical studies.
  • White to off-white solid form for ease of handling.
  • Limited solubility in DMSO (< 1 mg/mL).
  • Recommended storage: powder -20 °C (long term) or 4 °C (short term).
  • Product datasheet and SDS available for safety and handling.