Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile derivative (BI-2865) | 2937327-93-8 | 99.7% | 465.57 g/mol | C23H27N7O2S | 10 MG
SDP

Catalog No. 5000287166
Encompass_Preferred

BI-2865 is a non-covalent pan-KRAS small-molecule inhibitor used in preclinical research to block KRAS-driven signaling and assess antiproliferative effects in cellular and animal models. It is supplied as a high-purity solid for use in biochemical assays and in vivo studies.

  • Non-covalent pan-KRAS inhibitor active against multiple KRAS mutants.
  • Used for preclinical cellular and in vivo studies of KRAS signaling.
  • High purity (>99.6% by HPLC) suitable for biochemical assays.
  • Solid powder form; soluble in DMSO for stock solutions.
  • Molecular formula C23H27N7O2S; molecular weight 465.57 g/mol.
  • CAS number 2937327-93-8 for substance identification.

Catalog No. 50-002-87166 Supplier Medchemexpress LLC Supplier No. HY15372410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-2865 is a non-covalent pan-KRAS small-molecule inhibitor used in preclinical research to block KRAS-driven signaling and assess antiproliferative effects in cellular and animal models. It is supplied as a high-purity solid for use in biochemical assays and in vivo studies.

  • Non-covalent pan-KRAS inhibitor active against multiple KRAS mutants.
  • Used for preclinical cellular and in vivo studies of KRAS signaling.
  • High purity (>99.6% by HPLC) suitable for biochemical assays.
  • Solid powder form; soluble in DMSO for stock solutions.
  • Molecular formula C23H27N7O2S; molecular weight 465.57 g/mol.
  • CAS number 2937327-93-8 for substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.