Learn More
Cayman Chemical 4 5 7-TrImethoxyflavon 10mg
SDP
Catalog No. 502942243
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits AChE and BChE by 47.1 and 46.2%, respectively, in a cell-free assay at 0.1 mg/ml; scavenges radicals in a TEAC assay at 1 and 10 µM; inhibits RANKL-induced osteoclastic differentiation of RAW 264.7 cells at 10 µM; stimulates chloride secretion in Calu-3 cells
Catalog No. 50-294-2243 Supplier Cayman Chemical Supplier No. 4027110 MG
A flavonoid with diverse biological activities; inhibits AChE and BChE by 47.1 and 46.2%, respectively, in a cell-free assay at 0.1 mg/ml; scavenges radicals in a TEAC assay at 1 and 10 µM; inhibits RANKL-induced osteoclastic differentiation of RAW 264.7 cells at 10 µM; stimulates chloride secretion in Calu-3 cells