Learn More
Strem, An Ascensus Company CAS# 1101230-28-7. 250mg. 4,5-Bis-(di-i-propylphosphinomethyl)acridine, 98+%. MFCD18827624
SDP
Supplier:  Strem, An Ascensus Company 150415250MG
Encompass
CAS# 1101230-28-7. 250mg. 4,5-Bis-(di-i-propylphosphinomethyl)acridine, 98+%. MFCD18827624. Molecular Weight: 439.55. Molecular Formula: C27H39NP2. Color/form: yellow xtl. Strem# 15-0415. http://www.strem.com/catalog/v/15-0415/
Catalog No. 50-368-5