Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%

Catalog No. AAH2624103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2624103 1 g
AAH2624106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2624103 Supplier Thermo Scientific Chemicals Supplier No. H2624103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 161265-03-8 | C39H32OP2 | 578.63 g/mol

4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

Solubility
Soluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161265-03-8
Molecular Formula C39H32OP2
Molecular Weight (g/mol) 578.63
MDL Number MFCD00233866
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
PubChem CID 636044
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 228°C
Quantity 1 g
Solubility Information Soluble in organic solvents.
Formula Weight 578.62
Percent Purity 97%
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 605-249-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.