Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Bis(hydroxymethyl)imidazole 98.0+%, TCI America™
SDP

Catalog No. I04571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04571G 1 g
I04575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04571G Supplier TCI America Supplier No. I04571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33457-48-6
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00191661
InChI Key AGJXDKGTTMVHOU-UHFFFAOYSA-N
Synonym 1H-Imidazole-4,5-dimethanol
PubChem CID 573629
IUPAC Name [5-(hydroxymethyl)-1H-imidazol-4-yl]methanol
SMILES OCC1=C(CO)N=CN1

Specifications

Melting Point 162°C
Color White
Quantity 1 g
Formula Weight 128.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(hydroxymethyl)imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.