Learn More
Sigma Aldrich 4-(5-Chloro-2-methoxyphenyl)pyrimidin-2-amine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2,2-difluoro-2-(fluorosulfonyl)acetic acid reagent is employed as a difluorocarbene source for difluoromethylation of phenolic hydroxyl groups.2,2-difluoro-2-(fluorosulfonyl)acetic acid may be used in the following processes:
- Preparation of 1-difluoromethyl-2-oxo-1,2-dihydropyridine analogs by reacting with the corresponding 2-chloropyridines
- Prepration of silyl fluorosulfonyldifluoroacetate as new highly efficient difluorocarbene reagent for cyclopropanation of alkenes
- Regio- and stereoselective free radical fluoroalkylation of terminal alkenes and alkynes with iododifluoromethanesulfonamides
Specifications
Specifications
| Molecular Formula | C11H10ClN3O |
| Linear Formula | C11H10ClN3O |
| MDL Number | MFCD07381875 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 235.67 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.