Learn More
4',5'-Dibromofluorescein, Thermo Scientific™
Specifications
Specifications
| Melting Point | 270.0°C to 273.0°C |
| Loss on Drying | 5% max. |
| Infrared Spectrum | Authentic |
| Quantity | 25 g |
| Beilstein | 19,V,6,462 |
| Merck Index | 15,3027 |
| Solubility Information | (1% in hot 0.5 N NH4OH) Clear yellow to orange |
| Formula Weight | 490.11 |
| Chemical Name or Material | 4',5'-Dibromofluorescein |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 209-876- |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.