Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',5'-Dibromofluorescein, Thermo Scientific™

Catalog No. AC202431000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC202431000 100 g
AC202430050 5 g
AC202430250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC202431000 Supplier Thermo Scientific Chemicals Supplier No. 202431000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 596-03-2
Molecular Formula C20H10Br2O5
Molecular Weight (g/mol) 490.11
InChI Key ZDTNHRWWURISAA-UHFFFAOYSA-N
Synonym 4',5'-dibromofluorescein, solvent red 72, eosinic acid, japan orange 201, FD&C orange no. 5, c.i. solvent red 72, 4',5'-dibromofluororescein, 4,5-dibromofluorescein, d & c orange no. 5, japan orange no. 201
PubChem CID 11689
IUPAC Name 4',5'-dibromo-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)Br)OC5=C3C=CC(=C5Br)O

Specifications

Melting Point 270.0°C to 273.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Beilstein 19,V,6,462
Merck Index 15,3027
Solubility Information (1% in hot 0.5 N NH4OH) Clear yellow to orange
Formula Weight 490.11
Chemical Name or Material 4',5'-Dibromofluorescein
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 209-876-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.