Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dicarboxy-gamma-pentadecanolactone 98.0+%, TCI America™
SDP

Catalog No. D196325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D196325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D196325G Supplier TCI America Supplier No. D196325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 469-77-2
Molecular Formula C17H28O6
Molecular Weight (g/mol) 328.405
MDL Number MFCD00059059
InChI Key TUXHHVJPGQUPCF-SUMWQHHRSA-N
Synonym Spiculisporic Acid
PubChem CID 95228527
IUPAC Name (2R)-2-[(1R)-1-carboxyundecyl]-5-oxooxolane-2-carboxylic acid
SMILES CCCCCCCCCCC(C(=O)O)C1(CCC(=O)O1)C(=O)O

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 25 g
Formula Weight 328.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dicarboxy-gamma-pentadecanolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.