Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dichlorophthalonitrile 98.0+%, TCI America™
SDP

Catalog No. D218925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D218925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D218925G Supplier TCI America Supplier No. D218925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 139152-08-2
Molecular Formula C8H2Cl2N2
Molecular Weight (g/mol) 197.018
MDL Number MFCD00191408
InChI Key SRIJSZQFAMLVQV-UHFFFAOYSA-N
Synonym 1,2-Dichloro-4,5-dicyanobenzene
PubChem CID 853747
IUPAC Name 4,5-dichlorobenzene-1,2-dicarbonitrile
SMILES C1=C(C(=CC(=C1Cl)Cl)C#N)C#N

Specifications

Melting Point 183°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 197.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichlorophthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.