Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dicyano-1,3-dithiol-2-one 98.0+%, TCI America™
SDP

Catalog No. D32521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32521G Supplier TCI America Supplier No. D32521G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 934-31-6
Molecular Formula C5N2OS2
Molecular Weight (g/mol) 168.188
InChI Key BPDMXHRQYNWQSI-UHFFFAOYSA-N
Synonym 2-Oxo-1,3-dithiole-4,5-dicarbonitrile
PubChem CID 136741
IUPAC Name 2-oxo-1,3-dithiole-4,5-dicarbonitrile
SMILES C(#N)C1=C(SC(=O)S1)C#N

Specifications

Melting Point 124°C
Quantity 1 g
UN Number 3439
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dicyano-1,3-dithiol-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.