Learn More
Medchemexpress LLC 4,5-Dicyanoimidazole | 1122-28-7 | MFCD00005194 | 118.10 | 500 G
SDP
Catalog No. 5000445953
Encompass_Preferred
4,5-Dicyanoimidazole is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is for research use only and not for sale to patients.
  • Used in the activation of nucleotide phosphoramidites during solid-phase oligonucleotide synthesis
  • Synthesis of novel nucleotides
  • Often used as an alternative to tetrazole
Catalog No. 50-004-45953 Supplier Medchemexpress LLC Supplier No. HYW001962500G
4,5-Dicyanoimidazole is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. This product is for research use only and not for sale to patients.
  • Used in the activation of nucleotide phosphoramidites during solid-phase oligonucleotide synthesis
  • Synthesis of novel nucleotides
  • Often used as an alternative to tetrazole