Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,5-difluoro-2-methylbenzoic acid

Catalog No. CDS013819 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013819 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013819 Supplier Sigma Aldrich Supplier No. CDS013819250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-dimethylheptane is a branched saturated aliphatic hydrocarbon
  • It is one of the isomer of nonane
  • Henryαs law constant (solubility) of 2,3-dimethylheptane has been analyzed using characteristic and counting polynomials
  • Its standard heat of formation, combustion and vapourization at 25c has been calculated

Specifications

Molecular Formula C8H6F2O2
Linear Formula C8H6F2O2
Quantity 1 g
MDL Number MFCD08062377
Molecular Weight (g/mol) 172.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.