Learn More
Cayman Chemical 4 5-DIHDPA lacton 100ug
SDP
Catalog No. 502967609
Encompass_Preferred
A 1,4 cyclic ester derived from 4(5)-DiHDPA, which is produced by epoxidation of DHA at the α-4 double bond
Catalog No. 50-296-7609 Supplier Cayman Chemical Supplier No. 18344100 UG
A 1,4 cyclic ester derived from 4(5)-DiHDPA, which is produced by epoxidation of DHA at the α-4 double bond