Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dihydro-2-phenyloxazole 98.0+%, TCI America™
SDP

Catalog No. D21555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21555G 5 g
D215525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21555G Supplier TCI America Supplier No. D21555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7127-19-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00075541
InChI Key ZXTHWIZHGLNEPG-UHFFFAOYSA-N
Synonym (4,5-Dihydro-2-oxazolyl)benzene, 2-Phenyl(2-oxazoline)
PubChem CID 244030
IUPAC Name 2-phenyl-4,5-dihydro-1,3-oxazole
SMILES C1CN=C(O1)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 243°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4,5-Dihydro-2-phenyloxazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.