Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dihydroxy-1,3-benznedisulphonic acid disodium salt, 97.4%, MP Biomedicals™

Catalog No. ICN15774105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15774105 5 g
ICN15774180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15774105 Supplier MP Biomedicals, Inc Supplier No. 0215774105
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym tiron, tiferron, sodium catechol sulfate, disodium 4,5-dihydroxybenzene-1,3-disulfonate, 3,5-disulfocatechol disodium salt, sodium pyrocatechol-3,5-disulfonate, disodium pyrocatechol-3,5-disulfonate, sodium 1,2-dihydroxybenzenedisulfonate, 1,2-dihydroxybenzene-3,5-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid, 4,5-dihydroxy-, disodium salt
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Color Tan to White
Assay Percent Range 97.4%
Quantity 5 g
Formula Weight 332.22
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Dihydroxy-1, 3-benznedisulphonic acid disodium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.