Learn More
TARGETMOL CHEMICALS INC 4 5-Dihydroxyflavone 25MG
SDP
Catalog No. 502876810
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4',5-Dihydroxyflavone is a soybean LOX-1 inhibitor (Ki 102.6 uM) and a yeast alpha-Glucosidase inhibitor (IC50 66 uM). Purity 95.38%
Catalog No. 50-287-6810 Supplier TARGETMOL CHEMICALS INC Supplier No. T826025MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4',5-Dihydroxyflavone is a soybean LOX-1 inhibitor (Ki 102.6 uM) and a yeast alpha-Glucosidase inhibitor (IC50 66 uM). Purity 95.38%