Learn More
4,5-Dimethoxy-2-nitrobenzyl alcohol, 98%
Description
4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 144°C to 148°C |
| Quantity | 5 g |
| Beilstein | 1880093 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 213.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,5-Dimethoxy-2-nitrobenzyl alcohol |
Safety and Handling
| EINECSNumber | 213-806-4 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.