Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,5-Dimethoxy-2-nitrobenzyl bromide

Catalog No. 3928551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-285-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-285-51G Supplier Sigma Aldrich Supplier No. 3928551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,5-Dimethoxy-2-nitrobenzyl bromide

Specifications

CAS 53413-67-5
Melting Point 131°C to 133°C (lit.)
Molecular Formula C9H10BrNO4
Linear Formula O2NC6H2(OCH3)2 CH2Br
Quantity 1 g
MDL Number MFCD00192064
Synonym 6-Nitroveratryl bromide
Molecular Weight (g/mol) 276.08
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.