Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%

Catalog No. AC440770010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440770010 1 g
AC440770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440770010 Supplier Thermo Scientific Chemicals Supplier No. 440770010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 53413-67-5 | C9H10BrNO4 | 276.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53413-67-5
Molecular Formula C9H10BrNO4
Molecular Weight (g/mol) 276.09
InChI Key UEKFEYNZISYRRH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4,5-dimethoxy-2-nitrobenzene, 4,5-dimethoxy-2-nitrobenzyl bromide, 1-bromomethyl-2-nitro-4,5-dimethoxybenzene, bmndmb, benzene, 1-bromomethyl-4,5-dimethoxy-2-nitro, 6-nitroveratryl bromide, acmc-1ao6p, 4,5-dimenoxy-2-nitrobenzyl bromide, 2-nitro-4,5-dimethoxybenzyl bromide, 1-bromomethyl-4,5-dimethoxynitrobenzene
PubChem CID 3016812
IUPAC Name 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
SMILES COC1=C(C=C(C(=C1)CBr)[N+](=O)[O-])OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 132°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 276.09
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4, 5-Dimethoxy-2-nitrobenzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.