Learn More
Medchemexpress LLC 4,5-Dimethoxybenzene-1,2-diamine | 27841-33-4 | 97.89% | C8H12N2O2 | 100 MG
SDP
Catalog No. 5000435029
Encompass_Preferred
4,5-Dimethoxybenzene-1,2-diamine can be used as a standard for determining methyldiacetaldehyde, which is an intermediate product of glycolysis and a biomarker for diabetic ketoacidosis.
  • Used as a standard for determining methyldiacetaldehyde
  • Methyldiacetaldehyde is an intermediate product of glycolysis
  • Methyldiacetaldehyde is a biomarker for diabetic ketoacidosis
Catalog No. 50-004-35029 Supplier Medchemexpress LLC Supplier No. HY77873100MG
4,5-Dimethoxybenzene-1,2-diamine can be used as a standard for determining methyldiacetaldehyde, which is an intermediate product of glycolysis and a biomarker for diabetic ketoacidosis.
  • Used as a standard for determining methyldiacetaldehyde
  • Methyldiacetaldehyde is an intermediate product of glycolysis
  • Methyldiacetaldehyde is a biomarker for diabetic ketoacidosis