Learn More
Medchemexpress LLC 4,5-Dimethoxybenzene-1,2-diamine | 27841-33-4 | 97.9% | C8H12N2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY7787350MG
Encompass_Preferred
4,5-Dimethoxybenzene-1,2-diamine can be used as a standard for the determination of methyldiacetaldehyde, an intermediate product of glycolysis and a diabetic ketone biomarker of acidosis.
  • Can be used as a standard for the determination of methyldiacetaldehyde.
  • Functions as an intermediate product of glycolysis.
  • Serves as a diabetic ketone biomarker of acidosis.
Catalog No. 50-004-32747