Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,5-Dimethyl-o-phenylenediamine, 98%

Manufacturer:  Alfa Aesar™ H3269514

Quantity 25g

 View more versions of this product

Catalog No. AAH3269514


  Check Availability
Add to cart

Specifications

Description

Applications
4, 5-dimethyl-o-phenylenediamine used in the determination of aromatic aldehydes by high performance liquid chromatography after precolumn fluorescent derivatization. For the determination of vanillin.

Solubility
Slightly soluble in water.

Notes
Air & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, heat, light, acid chlorides, acid anhydrides.
Description & Specifications

Chemical Identifiers

3171-45-7
136.2
UN2811
4,5-Dimethyl-o-phenylenediamine
C8H12N2
MFCD00007729
4,5-Dimethyl-1,2-phenylenediamine; 4,5-Diamino-o-xylene

Specifications

3171-45-7
127° to 129°C
MFCD00007729
25g
4,5-Dimethyl-o-phenylenediamine
C8H12N2
136.2
UN2811
4,5-Dimethyl-1,2-phenylenediamine; 4,5-Diamino-o-xylene
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit