Learn More
Chemglass Life Sciences 4.5in Ground Flat Flange, Tube ID: 99mm, Tube OD: 105mm
SDP
Catalog No. CG13504
Encompass_Preferred
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 4.5in Ground Flat Flange, Tube ID: 99mm, Tube OD: 105mm.
Catalog No. CG13504 Supplier Chemglass Life Sciences Supplier No. CG13504
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 4.5in Ground Flat Flange, Tube ID: 99mm, Tube OD: 105mm.