Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(5-fluoro-1H-benzo[d]imidazol-2-yl)-N,N-dimethylaniline | 1458664-10-2 | MFCD20720876 | 99.6% | 255.29 | C15H14FN3 | 5 MG
SDP

Catalog No. 5000222052
Encompass_Preferred

β-catenin-IN-2 (Compound H1B1) is a small-molecule β-catenin inhibitor for research use, reported as a pink to red solid with high purity. It is characterized by molecular formula C15H14FN3 and a molecular weight of 255.29 g/mol, and is used in studies of colorectal cancer and β-catenin pathway biology.

  • Potent β-catenin inhibitor for colorectal cancer research.
  • Molecular formula C15H14FN3.
  • Molecular weight 255.29 g/mol.
  • Appearance: pink to red solid.
  • Purity around 99.6% by HPLC.
  • Available in small milligram-scale packages for laboratory studies.
  • Typically prepared and handled in solution (DMSO) for assays.

Catalog No. 50-002-22052 Supplier Medchemexpress LLC Supplier No. HY1364645MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-catenin-IN-2 (Compound H1B1) is a small-molecule β-catenin inhibitor for research use, reported as a pink to red solid with high purity. It is characterized by molecular formula C15H14FN3 and a molecular weight of 255.29 g/mol, and is used in studies of colorectal cancer and β-catenin pathway biology.

  • Potent β-catenin inhibitor for colorectal cancer research.
  • Molecular formula C15H14FN3.
  • Molecular weight 255.29 g/mol.
  • Appearance: pink to red solid.
  • Purity around 99.6% by HPLC.
  • Available in small milligram-scale packages for laboratory studies.
  • Typically prepared and handled in solution (DMSO) for assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.