Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, 4.5in Flange
SDP
Catalog No. CG13904
Encompass_Preferred
Clamp, Horseshoe Style, 4.5in Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. CG13904 Supplier Chemglass Life Sciences Supplier No. CG13904
Clamp, Horseshoe Style, 4.5in Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.