Learn More
Chemglass Life Sciences 4.5in O-Ring Flat Flange, Tube ID: 99mm, Tube OD: 105mm, O-Ring Size 348
SDP
Catalog No. CG13804
Encompass_Preferred
4.5in O-Ring Flat Flange, Tube ID: 99mm, Tube OD: 105mm, O-Ring Size 348. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.
Catalog No. CG13804 Supplier Chemglass Life Sciences Supplier No. CG13804
4.5in O-Ring Flat Flange, Tube ID: 99mm, Tube OD: 105mm, O-Ring Size 348. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.