Learn More
Sigma Aldrich 4-(5-methyl-1H-tetrazol-1-yl)aniline
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-(methylthio)propionaldehyde (methional) is the major product of chemical methionine conversion via strecker degradation in food.reactant involved in:
- Homolytic cross-coupling reactions
- Oxidative coupling and chemoselective oxidation
- Intermolecular alkyne hydroacylation reactions
- Oxidation of sulfides to sulfoxides
Specifications
Specifications
| Molecular Formula | C8H9N5 |
| Linear Formula | C8H9N5 |
| Quantity | 5 mL |
| MDL Number | MFCD00490014 |
| Molecular Weight (g/mol) | 175.19 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.