Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Methylenedithio-1,3-dithiole-2-thione 98.0+%, TCI America™
SDP

Catalog No. M11121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11121G Supplier TCI America Supplier No. M11121G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70800-59-8
Molecular Formula C4H2S5
Molecular Weight (g/mol) 210.36
MDL Number MFCD00137884
InChI Key NFKGILAHEQYFBG-UHFFFAOYSA-N
PubChem CID 5247527
IUPAC Name [1,3]dithiolo[4,5-d][1,3]dithiole-5-thione
SMILES C1SC2=C(S1)SC(=S)S2

Specifications

Melting Point 159°C
Quantity 1 g
Formula Weight 210.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Methylenedithio-1,3-dithiole-2-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.