Learn More
CHEMSCIENCE Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 4.5ML, Individually Serialized, Tolerance ± 0.01 mL
Supplier:  CHEMSCIENCE 123.302.04A
Encompass_Preferred
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 4.5ML, Individually Serialized, Tolerance ± 0.01 mL
Catalog No. NC2318865