Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(5-Trifluoromethyl-2-pyridyloxy)benzamidoxime, Thermo Scientific™

Catalog No. AAH50662MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50662MD 250 mg
AAH5066203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50662MD Supplier Thermo Scientific Chemicals Supplier No. H50662MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263161-34-8
Molecular Formula C13H10F3N3O2
Molecular Weight (g/mol) 297.237
MDL Number MFCD08060954
InChI Key RHAVFJMKHOLIST-UHFFFAOYSA-N
Synonym n-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxybenzenecarboximidamide, n-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzimidamide, z-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-carboximidamide, e-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzenecarboximidamide, n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzenecarboximidamide, 4-5-trifluoromethyl-2-pyridyloxy benzamidoxime, hydroxytrifluoromethylpyridinyloxybenzenecarboximidamide, z-n'-hydroxy-4-5-trifluoromethyl pyridin-2-yl oxy benzimidamide
PubChem CID 9582381
IUPAC Name N'-hydroxy-4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenecarboximidamide
SMILES C1=CC(=CC=C1C(=NO)N)OC2=NC=C(C=C2)C(F)(F)F

Specifications

Melting Point 160°C to 163°C
Quantity 250 mg
Formula Weight 297.24
Chemical Name or Material 4-(5-Trifluoromethyl-2-pyridyloxy)benzamidoxime
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.