Learn More
Medchemexpress LLC 4(5h)-thiazolone, 2-amino-5-[[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]methylene]-, (5z)- | 139340-56-0 | MFCD00906463 | 99.7% | 428.57 g·mol⁻¹ | C19H28N2O5S2 | 1 ML

Supplier: Medchemexpress LLC HY101438A10MM 1ML
Darbufelone mesylate is a research-grade small-molecule inhibitor that dual inhibits cellular PGF2α and LTB4 production and potently inhibits PGHS-2 (IC50 = 0.19 μM). Supplied as a solid and as a pre-made solution in DMSO, it is intended for research use only.
- Dual inhibition of PGF2α and LTB4 production.
- Potent PGHS-2 inhibitor (IC50 = 0.19 μM).
- Available as a solid and as a 10 mM solution in DMSO.
- High purity (99.65%).
- Soluble in DMSO (110 mg/mL); insoluble in water.
- Storage: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.