Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4(5H)-thiazolone, 2-amino-5-[[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]methylene] methanesulfonate | 139340-56-0 | MFCD00906463 | 99.7% | 428.57 g·mol⁻¹ | C19H28N2O5S2 | 10 MG
SDP

Catalog No. 5000249159
Encompass_Preferred

Darbufelone mesylate is a research-grade small molecule inhibitor that suppresses prostaglandin F2α and leukotriene B4 production by preferential inhibition of cyclooxygenase-2 (COX-2). Supplied as the mesylate salt for improved handling, it is intended for in vitro and in vivo research applications and is available as a solid or as a DMSO solution.

  • Potent COX-2 inhibition (IC50 ≈ 0.19 μM)
  • Dual inhibition of PGF2α and LTB4 production
  • High purity suitable for research (≈99.7%)
  • Soluble in DMSO (≈110 mg/mL), practically insoluble in water
  • Available as 10 mg solid or 10 mM solution in DMSO
  • Recommended storage sealed, away from moisture and light

Catalog No. 50-002-49159 Supplier Medchemexpress LLC Supplier No. HY101438A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Darbufelone mesylate is a research-grade small molecule inhibitor that suppresses prostaglandin F2α and leukotriene B4 production by preferential inhibition of cyclooxygenase-2 (COX-2). Supplied as the mesylate salt for improved handling, it is intended for in vitro and in vivo research applications and is available as a solid or as a DMSO solution.

  • Potent COX-2 inhibition (IC50 ≈ 0.19 μM)
  • Dual inhibition of PGF2α and LTB4 production
  • High purity suitable for research (≈99.7%)
  • Soluble in DMSO (≈110 mg/mL), practically insoluble in water
  • Available as 10 mg solid or 10 mM solution in DMSO
  • Recommended storage sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.