Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4(5H)-thiazolone, 2-amino-5-[(4-aminophenyl)methylene]-, (5Z)- | 1310744-67-2 | 99.0% | 100 MG
SDP

Catalog No. 5000232089
Encompass_Preferred

PFM39 is a small-molecule inhibitor of MRE11 exonuclease used for in-vitro research of DNA damage and repair pathways. It is supplied as a solid research reagent for biochemical and cell-based assays.

  • Selective MRE11 exonuclease inhibitor.
  • Molecular formula C10H9N3OS.
  • Molecular weight 219.26 g/mol.
  • Purity 99.0% (HPLC, as supplied).
  • Supplied as 100 MG solid; soluble in DMSO.
  • For research use only; not for therapeutic use.

Catalog No. 50-002-32089 Supplier Medchemexpress LLC Supplier No. HY120951100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PFM39 is a small-molecule inhibitor of MRE11 exonuclease used for in-vitro research of DNA damage and repair pathways. It is supplied as a solid research reagent for biochemical and cell-based assays.

  • Selective MRE11 exonuclease inhibitor.
  • Molecular formula C10H9N3OS.
  • Molecular weight 219.26 g/mol.
  • Purity 99.0% (HPLC, as supplied).
  • Supplied as 100 MG solid; soluble in DMSO.
  • For research use only; not for therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.