Learn More
BioSpec Products Probe, 0.45 cm dia
SDP
Catalog No. NC0433503
Encompass_Preferred
Replacement Tissue-Tearor probe, 0.45 cm dia size. Probe includes both the rotor and stator. Made of stainless steel with Teflon bushings.
Catalog No. NC0433503 Supplier BioSpec Products Supplier No. 9853G04
Replacement Tissue-Tearor probe, 0.45 cm dia size. Probe includes both the rotor and stator. Made of stainless steel with Teflon bushings.