Learn More
Medchemexpress LLC 4-[6-(4-isopropoxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline | 1206711-16-1 | 99.9% | 1 ML
SDP
Catalog No. 5000230673
Encompass_Preferred
DMH-1 is a selective small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors used in cell signaling and developmental biology research. It is supplied as a 10 mM solution in DMSO (1 mL) for in vitro assays.
  • Potent BMP type I receptor inhibitor; IC50s: ALK1 27 nM, ALK2 107.9 nM, ALK3 <5 nM, ALK6 47.6 nM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research (reported ~99.9% by HPLC).
  • Molecular formula C24H20N4O, molecular weight 380.44 g/mol.
  • Intended for cell-based and biochemical assays probing BMP signaling.
Catalog No. 50-002-30673 Supplier Medchemexpress LLC Supplier No. HY1227310MM 1ML
DMH-1 is a selective small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors used in cell signaling and developmental biology research. It is supplied as a 10 mM solution in DMSO (1 mL) for in vitro assays.
  • Potent BMP type I receptor inhibitor; IC50s: ALK1 27 nM, ALK2 107.9 nM, ALK3 <5 nM, ALK6 47.6 nM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research (reported ~99.9% by HPLC).
  • Molecular formula C24H20N4O, molecular weight 380.44 g/mol.
  • Intended for cell-based and biochemical assays probing BMP signaling.