Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[6-(4-propan-2-yloxyphenyl)pyrazolo[1,5-a]pyrimidin-3-yl]quinoline | 1206711-16-1 | >98.0% | 5 MG
SDP

Catalog No. 5000231466
Encompass_Preferred

DMH-1 is a small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors used in research to probe BMP/TGF-β signaling pathways. It provides potent, selective inhibition of ALK family kinases and is supplied as a high-purity analytical standard.

  • Selective BMP type I receptor inhibitor with low-nanomolar activity.
  • Reported IC50s: ALK1 27 nM; ALK2 107.9 nM; ALK3 <5 nM; ALK6 47.6 nM.
  • Molecular formula C24H20N4O and molecular weight 380.44 g/mol.
  • Purity ≥98% (HPLC).
  • CAS number 1206711-16-1 for substance identification.

Catalog No. 50-002-31466 Supplier Medchemexpress LLC Supplier No. HY122735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMH-1 is a small-molecule inhibitor of bone morphogenetic protein (BMP) type I receptors used in research to probe BMP/TGF-β signaling pathways. It provides potent, selective inhibition of ALK family kinases and is supplied as a high-purity analytical standard.

  • Selective BMP type I receptor inhibitor with low-nanomolar activity.
  • Reported IC50s: ALK1 27 nM; ALK2 107.9 nM; ALK3 <5 nM; ALK6 47.6 nM.
  • Molecular formula C24H20N4O and molecular weight 380.44 g/mol.
  • Purity ≥98% (HPLC).
  • CAS number 1206711-16-1 for substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.