Learn More
Medchemexpress LLC 4,6-bis(dimethylamino)-2-[(4-(4-(trifluoromethyl)benzoyl)amino)phenylmethyl]-5-pyrimidineacetic acid | 1093108-50-9 | ≥98.0% | 501.5 g/mol | C25H26F3N5O3 | 5 MG
SDP
Catalog No. 5000264577
Encompass_Preferred
BI-671800 is a small-molecule antagonist of the prostaglandin D2 receptor CRTH2/DP2 used in preclinical and clinical research on asthma and eosinophilic inflammation. It is provided as a crystalline solid (C25H26F3N5O3, 501.5 g/mol), is soluble in DMSO, and is reported with high purity and recommended low-temperature storage.
  • High purity (≥98.0%).
  • Validated antagonist activity at CRTH2/DP2 in biochemical and cell-based assays.
  • Suitable for in vitro and in vivo inflammation and respiratory research.
  • DMSO-soluble for straightforward formulation and dosing preparation.
  • Crystalline solid form for ease of handling and weighed dosing.
  • Recommended storage at -20 °C to maintain stability.
  • Well characterized: molecular formula C25H26F3N5O3; molecular weight 501.5 g/mol.
Catalog No. 50-002-64577 Supplier Medchemexpress LLC Supplier No. HY1141415MG
BI-671800 is a small-molecule antagonist of the prostaglandin D2 receptor CRTH2/DP2 used in preclinical and clinical research on asthma and eosinophilic inflammation. It is provided as a crystalline solid (C25H26F3N5O3, 501.5 g/mol), is soluble in DMSO, and is reported with high purity and recommended low-temperature storage.
  • High purity (≥98.0%).
  • Validated antagonist activity at CRTH2/DP2 in biochemical and cell-based assays.
  • Suitable for in vitro and in vivo inflammation and respiratory research.
  • DMSO-soluble for straightforward formulation and dosing preparation.
  • Crystalline solid form for ease of handling and weighed dosing.
  • Recommended storage at -20 °C to maintain stability.
  • Well characterized: molecular formula C25H26F3N5O3; molecular weight 501.5 g/mol.