Learn More
Strem, An Ascensus Company CAS# 261733-18-0. 500mg. 4,6-Bis(diphenylphosphino)phenoxazine, min. 98% NIXANTPHOS. MFCD03788937
SDP
Catalog No. 5090109871
Encompass
CAS# 261733-18-0. 500mg. 4,6-Bis(diphenylphosphino)phenoxazine, min. 98% NIXANTPHOS. MFCD03788937. Molecular Weight: 551.55. Molecular Formula: C36H27NOP2. Color/form: white to off-white pwdr. Strem# 15-0437. http://www.strem.com/catalog/v/15-0437/
Catalog No. 50-901-09871 Supplier Strem, An Ascensus Company Supplier No. 150437500MG
CAS# 261733-18-0. 500mg. 4,6-Bis(diphenylphosphino)phenoxazine, min. 98% NIXANTPHOS. MFCD03788937. Molecular Weight: 551.55. Molecular Formula: C36H27NOP2. Color/form: white to off-white pwdr. Strem# 15-0437. http://www.strem.com/catalog/v/15-0437/