Learn More
Medchemexpress LLC 4-[[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino]-N,N-diethylbenzamide | 444723-13-1 | MFCD09264153 | >98.0% | 422.5 g/mol | C23H30N6O2 | 50 MG
SDP
Catalog No. 5000239751
Encompass_Preferred
NU6140 is a purine-based small-molecule kinase inhibitor used in research to probe cell-cycle regulation and apoptotic responses. It is described as a selective CDK2-cyclin A inhibitor (reported IC50 ≈ 0.41 μM) that also inhibits Aurora A and Aurora B (reported IC50s 67 nM and 35 nM). The compound (CAS 444723-13-1; C23H30N6O2; MW 422.5 g/mol) is supplied as a high-purity reagent for in vitro and cell-based studies.
  • Selective CDK2-cyclin A inhibitor with reported IC50 ≈ 0.41 μM.
  • Potently inhibits Aurora A and Aurora B (reported IC50s 67 nM and 35 nM).
  • Suitable for in vitro and cell-based assays studying cell cycle and apoptosis.
  • Supplied as a high-purity research reagent (reported ≥98%).
  • Chemical formula C23H30N6O2 and molecular weight 422.5 g/mol.
  • Store under refrigerated conditions as recommended for laboratory reagents.
Catalog No. 50-002-39751 Supplier Medchemexpress LLC Supplier No. HY10741950MG
NU6140 is a purine-based small-molecule kinase inhibitor used in research to probe cell-cycle regulation and apoptotic responses. It is described as a selective CDK2-cyclin A inhibitor (reported IC50 ≈ 0.41 μM) that also inhibits Aurora A and Aurora B (reported IC50s 67 nM and 35 nM). The compound (CAS 444723-13-1; C23H30N6O2; MW 422.5 g/mol) is supplied as a high-purity reagent for in vitro and cell-based studies.
  • Selective CDK2-cyclin A inhibitor with reported IC50 ≈ 0.41 μM.
  • Potently inhibits Aurora A and Aurora B (reported IC50s 67 nM and 35 nM).
  • Suitable for in vitro and cell-based assays studying cell cycle and apoptosis.
  • Supplied as a high-purity research reagent (reported ≥98%).
  • Chemical formula C23H30N6O2 and molecular weight 422.5 g/mol.
  • Store under refrigerated conditions as recommended for laboratory reagents.