Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino]-N,N-diethylbenzamide | 444723-13-1 | MFCD09264153 | >98.0% | 422.52 g/mol | C23H30N6O2 | 25 MG
SDP

Catalog No. 5000239944
Encompass_Preferred

NU6140 is a small-molecule research compound that selectively inhibits cyclin-dependent kinase 2 (CDK2)-cyclin A (IC50 ≈ 0.41 μM) and shows reported activity against Aurora kinases. It is used in cell-cycle and apoptosis studies to probe kinase function in vitro. For laboratory research use only.

  • Selective CDK2-cyclin A inhibitor (IC50 ~0.41 μM).
  • Reported inhibition of Aurora A and Aurora B kinases.
  • Useful for inducing cell-cycle arrest and studying apoptosis pathways.
  • Molecular formula C23H30N6O2; molecular weight 422.52 g/mol.
  • High purity (>98.0%) suitable for research applications.
  • Soluble in DMSO and typically stored at -20 °C.
  • For laboratory research use only; not for human or clinical use.

Catalog No. 50-002-39944 Supplier Medchemexpress LLC Supplier No. HY10741925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NU6140 is a small-molecule research compound that selectively inhibits cyclin-dependent kinase 2 (CDK2)-cyclin A (IC50 ≈ 0.41 μM) and shows reported activity against Aurora kinases. It is used in cell-cycle and apoptosis studies to probe kinase function in vitro. For laboratory research use only.

  • Selective CDK2-cyclin A inhibitor (IC50 ~0.41 μM).
  • Reported inhibition of Aurora A and Aurora B kinases.
  • Useful for inducing cell-cycle arrest and studying apoptosis pathways.
  • Molecular formula C23H30N6O2; molecular weight 422.52 g/mol.
  • High purity (>98.0%) suitable for research applications.
  • Soluble in DMSO and typically stored at -20 °C.
  • For laboratory research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.