Learn More
Medchemexpress LLC 4-[[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino]-N,N-diethylbenzamide | 444723-13-1 | MFCD09264153 | 99.1% | 422.52 g/mol | C23H30N6O2 | 10 MG
SDP
Catalog No. 5000239705
Encompass_Preferred
NU6140 is a small-molecule research inhibitor described as a selective CDK2-cyclin A inhibitor that also inhibits Aurora A and Aurora B kinases. Supplied as a solid with reported high purity, it is used for cell-cycle and kinase research in both in vitro and in vivo experiments.
  • Selective CDK2-cyclin A inhibitor (IC50 0.41 μM).
  • Also inhibits Aurora A and Aurora B (IC50 67 nM and 35 nM).
  • High reported purity suitable for biochemical and cellular assays.
  • High DMSO solubility enables concentrated stock solutions.
  • Stable when stored as powder or in recommended solvent conditions.
  • Offered in small milligram pack sizes for early-stage research.
Catalog No. 50-002-39705 Supplier Medchemexpress LLC Supplier No. HY10741910MG
NU6140 is a small-molecule research inhibitor described as a selective CDK2-cyclin A inhibitor that also inhibits Aurora A and Aurora B kinases. Supplied as a solid with reported high purity, it is used for cell-cycle and kinase research in both in vitro and in vivo experiments.
  • Selective CDK2-cyclin A inhibitor (IC50 0.41 μM).
  • Also inhibits Aurora A and Aurora B (IC50 67 nM and 35 nM).
  • High reported purity suitable for biochemical and cellular assays.
  • High DMSO solubility enables concentrated stock solutions.
  • Stable when stored as powder or in recommended solvent conditions.
  • Offered in small milligram pack sizes for early-stage research.