Learn More
Medchemexpress LLC 4-[[6-(cyclohexylmethoxy)-9H-purin-2-yl]amino]-N,N-diethyl-benzamide | 444723-13-1 | ≥98.0% | 422.52 | C23H30N6O2 | 100 MG
SDP
Catalog No. 5000239840
Encompass_Preferred
NU6140 is a small-molecule research inhibitor selective for CDK2-cyclin A, used to probe cell-cycle regulation and kinase signaling. It also inhibits Aurora kinases and is provided as a research reagent with recommended cold-storage conditions.
  • Potent CDK2-cyclin A inhibition (IC50 0.41 μM).
  • Demonstrates 10-36-fold selectivity over other CDKs.
  • Inhibits Aurora A (IC50 67 nM) and Aurora B (IC50 35 nM).
  • Molecular weight 422.52; formula C23H30N6O2; CAS 444723-13-1.
  • Stable as powder under recommended cold storage for long-term use.
Catalog No. 50-002-39840 Supplier Medchemexpress LLC Supplier No. HY107419100MG
NU6140 is a small-molecule research inhibitor selective for CDK2-cyclin A, used to probe cell-cycle regulation and kinase signaling. It also inhibits Aurora kinases and is provided as a research reagent with recommended cold-storage conditions.
  • Potent CDK2-cyclin A inhibition (IC50 0.41 μM).
  • Demonstrates 10-36-fold selectivity over other CDKs.
  • Inhibits Aurora A (IC50 67 nM) and Aurora B (IC50 35 nM).
  • Molecular weight 422.52; formula C23H30N6O2; CAS 444723-13-1.
  • Stable as powder under recommended cold storage for long-term use.