Learn More
Medchemexpress LLC 4,6-diamino-5-formamidopyrimidine | 5122-36-1 | MFCD00047382 | 99.9% | 153.14 g/mol | C5H7N5O | 5 MG
SDP
Catalog No. 5000264239
Encompass_Preferred
FAPy-adenine is an oxidized DNA base and endogenous metabolite used as a biochemical standard and research reagent in studies of DNA oxidation and metabolic pathways. It has CAS 5122-36-1, formula C5H7N5O, and molecular weight 153.14 g/mol. For research use only; not for human therapeutic use.
  • High purity (99.9%).
  • Small pack sizes suitable for analytical and preparative work.
  • Soluble in DMSO (~17.86 mg/mL) for in vitro applications.
  • In vivo formulation guidance provided for common vehicles with measured solubility.
  • Stable as powder at -20 °C for up to 3 years.
  • Stable in solvent at -80 °C for up to 2 years.
Catalog No. 50-002-64239 Supplier Medchemexpress LLC Supplier No. HY1133035MG
FAPy-adenine is an oxidized DNA base and endogenous metabolite used as a biochemical standard and research reagent in studies of DNA oxidation and metabolic pathways. It has CAS 5122-36-1, formula C5H7N5O, and molecular weight 153.14 g/mol. For research use only; not for human therapeutic use.
  • High purity (99.9%).
  • Small pack sizes suitable for analytical and preparative work.
  • Soluble in DMSO (~17.86 mg/mL) for in vitro applications.
  • In vivo formulation guidance provided for common vehicles with measured solubility.
  • Stable as powder at -20 °C for up to 3 years.
  • Stable in solvent at -80 °C for up to 2 years.