Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,6-dichloro-N-methylnicotinamide-d3 | 1609531-34-1 | MFCD35118779 | 208.06 g/mol | C7H3D3Cl2N2O | 100 MG
SDP

Catalog No. 5000283866
Encompass_Preferred

4,6-Dichloro-N-methylnicotinamide-d3 is the deuterium-labeled analogue of 4,6-dichloro-N-methylnicotinamide intended for research applications as a tracer or internal standard in quantitative analytical methods.

  • Isotope labeled with three deuterium atoms (d3) for mass-shifted analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Chemical formula C7H3D3Cl2N2O and molecular weight 208.06 g/mol.
  • CAS number 1609531-34-1 for unambiguous identification.
  • Provided in a 100 mg quantity for laboratory use.
  • For research use only; refer to the Certificate of Analysis for purity and storage instructions.

Catalog No. 50-002-83866 Supplier Medchemexpress LLC Supplier No. HYW584477100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4,6-Dichloro-N-methylnicotinamide-d3 is the deuterium-labeled analogue of 4,6-dichloro-N-methylnicotinamide intended for research applications as a tracer or internal standard in quantitative analytical methods.

  • Isotope labeled with three deuterium atoms (d3) for mass-shifted analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Chemical formula C7H3D3Cl2N2O and molecular weight 208.06 g/mol.
  • CAS number 1609531-34-1 for unambiguous identification.
  • Provided in a 100 mg quantity for laboratory use.
  • For research use only; refer to the Certificate of Analysis for purity and storage instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.