Learn More
Accela Chembio Inc 4 | 6-dichloropyrimidine-5-carbaldehyde | 5g | 5305-40-8 | MFCD02257701 | 95+% | Shelf Life: 1620 Days | Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt
Supplier:  Accela Chembio Inc SY0026205G
Encompass_Preferred
4 | 6-dichloropyrimidine-5-carbaldehyde | 5g | 5305-40-8 | MFCD02257701 | 95+% | Shelf Life: 1620 Days | Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt
Catalog No. 50-353-467